Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Square Petri Dish, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70690-CS 70691 70691-CS
102097-176CS 1791.12 USD InStock
102097-176 102097-178 102091-156
Square Petri Dish, Electron Microscopy Sciences
Culture Dishes Petri Dishes
The square Petri dish with grid is consists of 36 of 13 x 13 mm squares, each 13 mm grid is marked numerically in one direction and alphabetically in the other
Made of polystyrene. Radiation sterile. This dish commonly referred to as a 100 x 15 mm dish. Free from optical distortion and sterile, these dishes are ideal for phase typing, susceptibility testing, plate counts, and probe assays.
The square Petri dish with grid is consists of 36 of 13 x 13 mm squares, each 13 mm grid is marked numerically in one direction and alphabetically in the other. This dish has four venting ribs into the underside of the lid to prevent condensation build-up.
This dish has four venting ribs into the underside of the lid to prevent condensation build-up.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty