Would you like to log in again?
Kodak T-MAX 100 Professional Films, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
KODAK®
74164 74172 74158 74162 74160 74173 74157
100488-102RL 487.74 USD InStock
100488-102 102091-272 102091-274 100488-104 100488-106 100492-126 100500-692
Kodak T-MAX 100 Professional Films, Electron Microscopy Sciences
Photographic Film
Very high degree of enlargement, expanded exposure latitude.
  • Medium speed (EI-100)
  • Extremely high sharpness, fine grain
  • Total darkness
Especially useful for detailed subjects when maximum image quality is needed. Recommended for copying black and white photographs.
Recommended kodak developers: T-Max, D-76, HC-110, Microdol-X, Duraflo RT.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty