Would you like to log in again?
Standard Pin Stub SEM Mount Small Non-Slotted Head, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
75186
102091-298PK 406.23 USD InStock
102091-298
Standard Pin Stub SEM Mount Small Non-Slotted Head, Electron Microscopy Sciences
Specimen Mounting Supplies
Non-Slotted head pin stub mounts are for use with Cambridge, Leo, Philips 500, Cameca SEMs. Made from lathe finished ultra-pure aluminium.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty