Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
3M™ Double Sided Tape Scotch 665 or Scotch 666, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77115 77102 77116 77101 77118
102091-328EA 43.74 USD InStock
102091-328 102680-464 102091-324 100500-704 100492-314
3M™ Double Sided Tape Scotch 665 or Scotch 666, Electron Microscopy Sciences
Tapes
Non-conductive transparent tape with adhesive on both sides. Available with or without white paper liner in different widths.
  • Good adhesion and cleanliness
  • Versions with liner: white paper as liner is slit in the center to facilitate its removal
  • 77 mm (3") core
32.9 m (36 yds.) long
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty