Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
EMS Lanthanum and Cerium Hexaboride Cathodes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
80929-L-CM 80933-L 80930 80920 80922-16-L 80921-L 80929-L-20 80930-L 80928 80929-20 80926-S1 80923-L 80921 80927-L 80922-20-L 80928-L 80929-20S 80929-L-P 80928-L-S 80931 80922-20 80926-L-S1 80920-L 80926-L-T1 80933 80929-L-20S 80922-16 80923 80929-CM 80926-L-T 80926-S2 80926-L-S2 80927-L-HT 80926-T 80931-L 80927-HT 80927 80929-P
102099-358EA 1972.88 USD InStock
102099-358 102091-360 102099-336 102099-314 102099-312 102099-334 102091-362 102099-356 102099-354 102099-310 102099-332 102099-330 102099-352 102099-318 102099-338 102091-366 102099-316 102099-340 102099-348 102099-304 102099-326 102091-350 102099-302 102099-346 102099-324 102099-344 102099-322 102099-300 102099-320 102099-342 102091-356 102091-358 102091-352 102099-308 102099-306 102091-354 102099-328 102099-350
EMS Lanthanum and Cerium Hexaboride Cathodes, Electron Microscopy Sciences
Lights Hollow Cathode Lamps
For applications with large beam spot sizes, where large total current and current density are required, large, flat crystal faces of LaB6 or CeB6 can be the cathodes of choice.
  • Excessive operating temperatures accelerate evaporation
This regime is unsuitable for point sources such as field emitters, which are unable to provide sufficient total current, and has been thought of as the realm of the dispenser cathode. However, LaB6 and CeB6 may be more suitable, being particularly robust and resistant to chemical poisoning. They have modest vacuum requirements and long shelf life, and need only be brought up to operating temperature to provide emission, eliminating the activation procedure required of dispenser cathodes.
This decreases the life of the cathode. Care must be taken to properly optimize cathode temperature to obtain the required emission without overheating the crystal. CeB6 has another advantage over LaB6 relating to lifetime: its evaporation rate at normal operating temperatures near 1800 K is lower than that of LaB6. So long as care is taken to operate the cathode below 1850 K, CeB6 should maintain an optimum tip shape longer, and therefore last longer.
These trends allow us to tailor our cathodes to the requirements of practically all thermionic cathode applications. For example, SEM and most transmission electron microscope (TEM) applications are best served by a 90° cone angle and a 16 mm flat tip. This combination provides high brightness, a moderate source size, and very good lifetime. High resolution TEMs require a 60° cone and a 5 mm flat tip for very high brightness and a small source size. In applications requiring high total current in a large beam spot, a 310 oriented crystal in a "top hat" configuration may be preferred, providing a slightly lower work function and large emitting surface. We excel at developing specialized cathodes for custom applications and research purposes. Contact us for your custom cathode needs.
The structure of the MVM is amazingly robust, sustaining reasonable impact without deviating from structural specifications. Because the graphite pads shield evaporation of the crystal in the direction of the clamping force, the emitter crystal can be fully utilized without degradation of the mount. Structural failure of the MVM is not a concern when the cathode is operated within the correct temperature and pressure range. Typically, the beam stability of the Mini Vogel Mount cathode exceeds the specifications of the system in which it runs.
They can provide long-term, stable operation at current densities up to 50 A/cm2, and may be fabricated in a variety of shapes and with many different heating and mounting configurations. The design of the cathode tip is critical for maximum lifetime and optimum performance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty