Would you like to log in again?
Kodak Ektatherm XLS Media Products, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
89058 89050 89059 89057 89056 89052 89053
102091-382PK 223.64 USD InStock
102091-382 102091-370 102091-384 102091-380 102091-378 102091-374 102091-376
Kodak Ektatherm XLS Media Products, Electron Microscopy Sciences
Film Processing Supplies
We are offering a full range of media products for outstanding image stability and near-photographic quality continuous tone thermal prints. Our media products feature excellent color reproduction, including superior flesh tones and durability. The unique Kodak XtraLife Ribbon overcoats prints with a rugged transparent layer which prevents destructive dye transfer as well as it resists fingerprints and fading. NOTE: Only laminates to paper. When printing transparency material with the XTRALIFE Ribbons the laminate patch is bypassed. For use with the 48 MB Printer only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty