Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Edwards RV8 Vacuum Pump, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
91025 91025-E
100495-462EA 6235.52 USD InStock
100495-462 102091-400
Edwards RV8 Vacuum Pump, Electron Microscopy Sciences
Pumps Vacuum Pumps
Rotary vacuum pump; Double stage. Ultra Grade 19 oil is recommended for use with the RV8. Weight 50 lbs (24 kg).

  • 6.9 cfm (195 L/min) displacement
  • Ultimate vacuum (without gas ballast) 1.5x10-3 torr
  • Max. inlet water pressure vapor: 29 torr
  • Max. water vapor pumping rate: .48 lb/h
  • Motor: 3/4 hp
  • Oil capacity Max/Min 0.75L/0.43

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty