Would you like to log in again?
Dumont Tweezers Style N5AC, Dumont Switzerland, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72856-DZ 72856-D 0202-N5AC-PO 0207-N5ACTI-PO
100499-820DZ 1109.17 USD InStock
100499-820 102091-570 76375-934 100488-320
Dumont Tweezers Style N5AC, Dumont Switzerland, Electron Microscopy Sciences
Forceps
Anti-capillary, type 5, super thin tips.
  • Hand made of the highest quality steels and alloys
  • High precision Swiss manufacturing
  • Perfectly finished to suit your application
Inox 02 is a standard magnetic stainless steel composed of C, Mn, Cr and Si. This alloy is not as hard as carbon steels because it contains Chromium. Lnox 02 will resist temperatures of around 400°C (DIN 50 914), but cannot be sterilized.
Dumoxel is composed of C, Cr, Ni, Mo and Cu. This alloy was developed and patented by Dumont to offer the best in resistance to corrosion. Dumoxel also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel is 95% antimagnetic, resistant up to temperatures of around 400 °C (DIN 50 914) and is suitable for autoclave sterilization at 270 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty