Would you like to log in again?
Dumont Tweezers Style 4AC, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0208-4AC-PO
102091-600EA 96.58 USD InStock
102091-600
Dumont Tweezers Style 4AC, Electron Microscopy Sciences
Forceps
Anti-capillary, very thin tips. Length: 110 mm.
Inox 08 is a medical stainless steel composed of C, Mn, Cr, Mo and V that provides an excellent resistance to corrosion and a good resistance to salt. Although not as hard as Carbon steels, Inox 08 offers excellent resistance to corrosion. This magnetic alloy supports temperatures of around 400°C (DIN 50914) and is suitable for autoclave sterilization at 180°C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty