Would you like to log in again?
Dumont Tweezers Style 0, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0304-0-PO-1 0302-0-PO 0304-0-CO
76499-470EA 31.5 USD InStock
76499-470 102091-664 102091-622
Dumont Tweezers Style 0, Electron Microscopy Sciences
Forceps
Antimagnetic is composed of C, Cr, Ni and Mo
Geneva pattern, thin tips. Length: 115 mm.
Inox 02 is a standard magnetic stainless steel composed of C, Mn, Cr and Si. This alloy is not as hard as carbon steels because it contains Chromium. lnox 02 will resist temperatures of around 400°C (DIN 50 914), but cannot be sterilized.
Antimagnetic is composed of C, Cr, Ni and Mo. It offers a good resistance to corrosion thanks to its high concentration of molybdenum and is 80% antimagnetic but is not as hard as Inox. Antimagnetic is resistant up to temperatures of around 400°C (DIN 50 914) and is suitable for autoclave sterilization at 270°C.
It offers a good resistance to corrosion thanks to its high concentration of molybdenum and is 80% antimagnetic but is not as hard as Inox. Antimagnetic is resistant up to temperatures of around 400°C (DIN 50 914) and is suitable for autoclave sterilization at 270°C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty