Would you like to log in again?
Dumont Tweezers Style 5, Dumont Switzerland, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0302-5-PO 0304-5-TF 72702-DTIP-DZ 72702-DTIP 0303-5-PO 0303-5-TF 0302-5-CO
102773-132PK 806.51 USD InStock
102773-132 76375-960 102091-636 102091-698 102091-654 102091-656 102091-634
Dumont Tweezers Style 5, Dumont Switzerland, Electron Microscopy Sciences
Forceps
Swiss made precision tweezers from Dumont.
  • Hand made of the highest quality steels and alloys
  • High precision Swiss manufacturing
  • Perfectly finished to suit your application
Style 5 tweezers have super thin tips
Dumoxel is composed of C, Cr, Ni, Mo and Cu and offers the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel is 95% antimagnetic, resistant up to temperatures of around 400 ° C (DIN 50 914) and is suitable for autoclave sterilization at 270 ° C.
Length 110 mm. Tip measures 0.10 mm W x 0.06 mm thick.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty