Would you like to log in again?
Dumont Tweezers Style N2A, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0304-N2A-PO
102091-710EA 44.78 USD InStock
102091-710
Dumont Tweezers Style N2A, Electron Microscopy Sciences
Forceps
Type 2A, round tips, 118 mm.
Antimagnetic is composed of C, Cr, Ni and Mo. It offers a good resistance to corrosion thanks to its high concentration of molybdenum and is 80% antimagnetic but is not as hard as Inox. Antimagnetic is resistant up to temperatures of around 400°C (DIN 50 914) and is suitable for autoclave sterilization at 270°C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty