Would you like to log in again?
Dumont Tweezers Style 11, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0305-11NI-CO 0305-11NI-PO
102091-728EA 44.78 USD InStock
102091-728 102091-730
Dumont Tweezers Style 11, Electron Microscopy Sciences
Forceps
Strong tips. Ideal to manipulate delicate pieces. Length: 120 mm. Nickel is a 100% antimagnetic alloy composed of Cu, Ni, Mn and Zn. lt is widely appreciated as it does not damage the components handled by it.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty