Would you like to log in again?
Mirskys Fixative, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
15561-20 15561-10 15561-05 15560
102091-834EA 70.81 USD InStock
102091-834 102091-836 102091-832 102090-982
Mirskys Fixative, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Mirsky’s Fixative™ is a superior fixing agent for use in immunohistological and immunological protocols
  • Saponin/Glyoxylate Fixative
  • Preserves Immunohistological activity
  • Contains no formaldehyde
  • Fast-acting
Double reactive sites afford excellent cross-linking properties while maintaining sample enzyme activity. Samples processed in Mirsky’s Fixative for light microscopy can subsequently be used in electron microscopy procedures. Special buffer systems may be used in place of the buffer provided.
Use as replacement for formalin and/or glutaraldehyde fixatives in immunohistological and immnocytological staining protocols. Tissue size is unlimited, as long as the sample is no thicker than 0.5cm in at least one plane to assure uniformity of tissue penetration. The gross visual appearance will be different than in formaldehyde and tissue may seem ""raw"". This is due to double site binding of the fixative with a resultant reduction in tissue shrinkage. Microscopic examination of tissue morphology will be noticeably improved. This material is intended to maintain enzyme and antibody activity. For best results, tissue section of high digestive enzyme content should be thoroughly rinsed in saline solution before fixation (e.g. trypsin in intestinal samples). Once fixed, tissue may be retained in Mirsky’s Fixative and ethanol indefinitely.
Mirsky’s Fixative is normally distributed as a concentrated two bottle system, although high through-put laboratories often use the single bottle ready-to-use format. The two bottle system is comprised of Mirsky’s Fixative 10X Concentrated and Mirsky’s Fixative 10X Buffer. To reconstitute to working strength, add 1 part Mirsky’s Fixative 10X Buffer to 8 parts distilled or deionized water, mix thoroughly, then add 1 part Mirsky’s Fixative 10X Concentrate and mix again.
The two bottle system has a shelf life of 12 months while the ready-to-use format has a shel life of 30 days.
Mirsky’s contains no formaldehyde or glutaraldehyde. Therefore, it has considerably reduced toxicity and virtually no odor. Mirsky’s Fixative is neutral, buffered, and isotonic (308 mOsmol). Additionally, Mirsky’s Fixative does not contain toxic or hazardous buffers such as cacodylate or barbital. Hardening or shrinkage of tissue is considerably reduced.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty