Would you like to log in again?
Carson's Phosphate Buffered Formalin, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
15732-10-S 15732-05
102091-844EA 63.49 USD InStock
102091-844 102091-840
Carson's Phosphate Buffered Formalin, Electron Microscopy Science
Stains Microscopy and staining reagents
Millonig’s phosphate buffered formalin as modified by Carson, Martin and Lynn, by using 4% formalin in 0.1M sodium phosphate monobasic with sodium hydroxide to pH 7.2
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty