Would you like to log in again?
Carsons Phosphate Buffered Paraformaldehyde, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
15733-10 15733-05
102091-848EA 66.54 USD InStock
102091-848 102091-846
Carsons Phosphate Buffered Paraformaldehyde, Electron Microscopy Science
Stains Microscopy and staining reagents
For some tissues, the use of formalin (contains 10 – 15% Methanol) is not suitable, we offer the alternative formaldehyde (prepared from paraformaldehyde – contains no methanol) for your convenience.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty