Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
CoreDish™ Multiple Biopsy Containers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
15736-50 15736-20 15736-30 15736-10 15736-40
102091-900PK 113.12 USD InStock
102091-900 102091-902 102091-896 102091-898 102091-894
CoreDish™ Multiple Biopsy Containers, Electron Microscopy Sciences
Embedding Cassettes Biopsy Processing Cassettes
We are proud to offer a multi-compartment container in the shape of a dish and half prefilled with 10% Neutral buffered Formalin, for holding and transporting biopsies
It is supplied with a leak-proof closure with O-ring ensuring total protection of contents. It conforms to OSHA directives. These CoreDisn™ measure only 15 x 95 mm. Each compartment is clearly identified to allow proper placement and visualization of the biopsy being inserted. Thanks to CoreDish™ it is no more necessary to use a multiple of individual containers, thereby reducing cost and risks of confusion. Few recommendations concerning how the biopsies should be handled have been published. Performing a large number of biopsies means an increase in the number of containers handled and consequently a technical overload of the transmission network, which occurs without any financial counterpart. A new approach had to be developed in order to increase productivity.
We are proud to offer a multi-compartment container in the shape of a dish and half prefilled with 10% Neutral buffered Formalin, for holding and transporting biopsies. It is supplied with a leak-proof closure with O-ring ensuring total protection of contents. It conforms to OSHA directives. These CoreDisn™ measure only 15 x 95 mm. Each compartment is clearly identified to allow proper placement and visualization of the biopsy being inserted. Thanks to CoreDish™ it is no more necessary to use a multiple of individual containers, thereby reducing cost and risks of confusion.
CoreDish™ available in many configurations to serve in different biopsy, such as prostate, upper GI tract and lower GI tract. A label allows recording the related information, patient ID, doctor, date and time.
M970-D5B-2 CoreDish™ Breast Biopsy Container is half prefilled with 10% Formalin. This multi-compartment container (out of five compartments, four are labeled: LUQ, RUQ, LLQ, RLQ) in the shape of a dish and half prefilled with 10% Neutral Buffered Formalin, for holding and transporting biopsies. It is supplied with a leak-proof screw closure with O-ring ensuring total protection of contents. It conforms to OSHA directives. The dish measures only 15 x 95 mm in diameter. Each compartment is clearly identified to allow proper placement and visualization of the breast biopsy being inserted. A writing area for patient information is provided.
M970-D8P CoreDish™ Prostate Biopsy Container is half prefilled with 10% Formalin. For prostate biopsies. Eight compratments. Leakproof seal with O-ring lid. An area for patient information is provided. Seven labeled com partments: Base, Lateral Base, Medium, Lateral Medium, Apex, Lateral Apex.
M970-D8UGI CoreDish™ Upper GI Biopsy Container is half prefilled with 10% Formalin. For upper GI track biopsies. Eight compratments. Leakproof seal with O-ring lid. An area for patient information is provided. Seven labeled compartments: Gastric Card, Gastric Body, GE Junction, Gastric ATR, Distal Esophage, Pylorus, Duodeum.
M970-D8LGI CoreDish™ Lower Gi Biopsy Container is half prefilled with 10% Formalin. For lower GI track biopsies. Twelve compratments. Leakproof seal with O-ring lid. An area for patient information is provided. Ten labeled compartments: Proximal Flexure Colon, Hepatic Fleure Colon, Distal Transverse Colon, Ascending Colon, Splenic flexure Colon, Cecum, Decending Colon, Terminal lleum, Rectum, Sigmmoid Colon.
M970-D12P CoreDish™ Prostate Biopsy Container is half prefilled with 10% Formalin. For prostate biopsies. Twelve compratments. Leakproof seal with O-ring lid. An area for patient information is provided. TWelve labeled compartments: L Base, R Base, L Lateral Base, R Lateral Base, L Medium, R Medium, L Lateral Medium, R Lateral Medium, L Lateral , L Apex, R Apex, R Lateral Apex. CoreDish™ available in many configurations to serve in different biopsy, such as prostate, upper GI tract and lower GI tract. A label allows recording the related information, patient ID, doctor, date and time.
M970-D5B-2 CoreDish™ Breast Biopsy Container is half prefilled with 10% Formalin. This multi-compartment container (out of five compartments, four are labeled: LUQ, RUQ, LLQ, RLQ) in the shape of a dish and half prefilled with 10% Neutral Buffered Formalin, for holding and transporting biopsies. It is supplied with a leak-proof screw closure with O-ring ensuring total protection of contents. It conforms to OSHA directives. The dish measures only 15 x 95 mm in diameter. Each compartment is clearly identified to allow proper placement and visualization of the breast biopsy being inserted. A writing area for patient information is provided.
M970-D8P CoreDish™ Prostate Biopsy Container is half prefilled with 10% Formalin. For prostate biopsies. Eight compratments. Leakproof seal with O-ring lid. An area for patient information is provided. Seven labeled com partments: Base, Lateral Base, Medium, Lateral Medium, Apex, Lateral Apex.
M970-D8UGI CoreDish™ Upper GI Biopsy Container is half prefilled with 10% Formalin. For upper GI track biopsies. Eight compratments. Leakproof seal with O-ring lid. An area for patient information is provided. Seven labeled compartments: Gastric Card, Gastric Body, GE Junction, Gastric ATR, Distal Esophage, Pylorus, Duodeum.
M970-D8LGI CoreDish™ Lower Gi Biopsy Container is half prefilled with 10% Formalin. For lower GI track biopsies. Twelve compratments. Leakproof seal with O-ring lid. An area for patient information is provided. Ten labeled compartments: Proximal Flexure Colon, Hepatic Fleure Colon, Distal Transverse Colon, Ascending Colon, Splenic flexure Colon, Cecum, Decending Colon, Terminal lleum, Rectum, Sigmmoid Colon.
M970-D12P CoreDish™ Prostate Biopsy Container is half prefilled with 10% Formalin. For prostate biopsies. Twelve compratments. Leakproof seal with O-ring lid. An area for patient information is provided. TWelve labeled compartments: L Base, R Base, L Lateral Base, R Lateral Base, L Medium, R Medium, L Lateral Medium, R Lateral Medium, L Lateral , L Apex, R Apex, R Lateral Apex.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty