Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formaldehyde 4% in aqueous solution, EM Grade

Supplier: Electron Microscopy Sciences
Danger
Synonyms: MethylaldehydeFormolFormalinParaformaldehyde aqueous solution

157-4 157-4-100
102091-904PK 57.35 USD InStock
102091-904 102091-906
Formaldehyde 4% in aqueous solution, EM Grade
Formaldehyde
Paraformaldehyde - methanol free solution.

A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives.

Since the DOT (Department of Transportation) has changed the definition of what a non dangerous solution of formaldehyde is to under 10%, we are now offering an 8% and a 4% solution. This will allow us to ship overnight as well as overseas without any hazardous charges saving our customers a good deal of money.

Packaging: Easy to break, prescored, 10 ml ampoules sealed under inert gas, or 100 ml bottles.


Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/71/28984871.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty