Would you like to log in again?
Fixation Buffers, Glutaraldehyde, Pre-Made, Ready-to-Use, in Buffer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
16536-15 16539-15 16538-10 16539-07 16536-08 16539-65 16537-10 16537-18 16537-15 16539-30 16539-60 16539-40 16538-08 16537-07 16536-20 16539-50 16539-06 16537-19 16539-70 16536-07 16537-20-S 16539-55 16537-21 16538-15 16538-07 16539-45 16537-06 16535-05 16536-05 16537-16 16537-20 16539-35 16538-20 16539-08 16537-17 16538-06 16539-25 16537-08 16538-05 16535-10 16539-20 16536-10 16536-06 16537-05
102092-008EA 561.83 USD InStock
102092-008 102092-070 102092-030 102092-074 102092-052 102091-998 102092-050 102092-072 102092-056 102092-078 102092-012 102092-034 102092-010 102092-054 102092-032 102092-076 102092-038 102092-016 102092-036 102092-058 102092-014 102092-018 102091-996 102092-060 102092-082 102092-080 102092-042 102092-064 102092-020 102092-040 102092-062 102092-024 102092-046 102092-068 102092-002 102092-022 102092-066 102092-044 102092-000 102092-006 102092-028 102092-048 102092-004 102092-026
Fixation Buffers, Glutaraldehyde, Pre-Made, Ready-to-Use, in Buffer, Electron Microscopy Sciences
Buffers Microscopy Buffers
Flow Cytometry Staining Buffer is designed for use in immunofluorescent staining protocols of cells in suspension.
This Flow Cytometry Staining Buffer is a buffered saline solution containing fetal bovine serum and sodium azide (0.09%) as a preservative. This buffer can be used for antibody and cell dilution steps, as well as all the wash steps required for the surface staining and flow cytometric analysis. This buffer contains animal serum proteins to help minimizing non-specific binding of antibodies.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty