Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Fluorescence Microscopy Immersion Oil, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
16916-04 16916-01 16919-16 16919 16915-04 16919-01
102090-990EA 30.96 USD InStock
102090-990 102092-096 100501-602 102092-094 100496-524 100496-526
Fluorescence Microscopy Immersion Oil, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Two new PCB-free immersion oils are availablefor fluorescence microscopy.
  • FF has virtually no background fluorescence
  • Extremely low fluorescence by LDF
  • Low fluorescence by HF
FF has virtually no background fluorescence
Extremely low fluorescence by LDF
Low fluorescence by HF
New Immersion oil HF provide low fluorescence and the correct optical properties and FF has irtually no fluorescence but slightly less than ideal optical properties. Applicator bottles have a glass ball rod in the cap.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty