Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Fluoro-Gel Substitute for BioMeda™ Gel-Mount, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Fluoro-Gel
17985-11 17985-31 17985-61 17985-41 17985-50 17985-30 17985-40 17985-10 17985-51 17985-60
102092-140EA 146.19 USD InStock
102092-140 102092-150 102092-144 102092-122 102092-142 102092-138 102092-126 102092-148 102092-136 102092-146
Fluoro-Gel Substitute for BioMeda™ Gel-Mount, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Fluoro-Gel is a water based mounting medium designed for the permanent mounting of fluorescent stained tissues, which may either be damaged or soluble in organic solvents, such as xylene or toluene. This unique formula prevents rapid photo-bleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa Fluoro 488, Alexa Fluoro 594, Green Fluorescent Protein (GFP), Tetramethylyrhodamine, Redox, Phycoerythrin (RP-E), Phyocyanin (PC), and Allophycocyanin (APC). The fluorescence is retained during prolonged storage at 4 °C in the dark. Application: Immunofluorescence, confocal microscope.
Certifications: Certificates of analysis are available from Electron Microscopy Sciences. Please contact us or visit our website to search by lot or product number.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty