Would you like to log in again?
Omnimount, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
Omnimount™
17997-01
102092-152EA 80.37 USD InStock
102092-152
Omnimount, Electron Microscopy Science
Stains Microscopy and staining reagents
Specially developed to provide compatibility with National Diagnostic clearing agents, HistoClear II. Omnimount™ combined outstanding optical characteristics with low fluororescence and exceptional durability.
  • Optical clarity combined with exceptional durability
  • Compatible with HistoClear II
  • Higher flash point and lower toxicity than xylene based mountants
In addition to being the ideal partner for HistoClear II, Omnimount is a truly universal mounting medium compatible with all common clearing agents: xylene, toluene, limonene, and petroleum derived products.
The Omnimount solvent has a higher flash point and a lower toxicity than xylene based mountants, so Omnimount™ provides both a safer work environment and reduce shipping costs.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty