Would you like to log in again?
Histosol™ Xylene Substitute, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Histosol™
23405-01 23405-04
102092-290EA 159.97 USD InStock
102092-290 102092-292
Histosol™ Xylene Substitute, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Histosol™ is the original high flash point (114°F TCC) histological clearing agent.
It is intended to be used as a replacement for xylene where the hazards associated with aromatic hydrocarbon vapors are to be reduced. Museum-quality tissue slides can be prepared with Hitosol without change in protocol or procedure. Histosol is manufactured from petrochemical products and is miscible in all proportions with ethanol, isopropanol, and t-butanol. It is also miscible with all paraffin-based tissue embedding media and all permanent mounting materials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty