Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

Molybdenum, disc, drilled apertures 3 mm O.D., Thickness 0.12 mm

Supplier: Electron Microscopy Sciences

43060-MO 43150-MO 43200-MO 43010-MO 43300-MO 43100-MO 43020-MO 43050-MO 43030-MO 43070-MO
102092-626EA 158.83 USD InStock
102092-626 102092-636 102092-624 102092-634 102092-628 102092-638 102092-640 102092-622 102092-632 102092-630
Molybdenum, disc, drilled apertures 3 mm O.D., Thickness 0.12 mm
Molybdenum
Hole Sizes not listed are available upon special request.


Formula: Mo
MW: 95.94 g/mol
MDL Number: MFCD00003465
CAS Number: 7439-98-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty