Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Molybdenum, disc, drilled apertures 3 mm O.D., Thickness 0.24 mm

Supplier: Electron Microscopy Sciences

44030-MO 44100-MO 44005-MO 44010-MO 44200-MO 44300-MO 44060-MO 44020-MO 44025-MO 44050-MO 44015-MO 44070-MO 44400-MO 44040-MO 44150-MO
102092-648EA 155.69 USD InStock
102092-648 102092-658 102092-668 102092-646 102092-656 102092-662 102092-650 102092-660 102092-670 102092-644 102092-666 102092-654 102092-642 102092-664 102092-652
Molybdenum, disc, drilled apertures 3 mm O.D., Thickness 0.24 mm
Molybdenum
Hole Sizes not listed are available upon special request.


Formula: Mo
MW: 95.94 g/mol
MDL Number: MFCD00003465
CAS Number: 7439-98-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty