Would you like to log in again?
MicroMounts™ Crystal Manipulation Tool, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60122-S 60117-25 60125-25 60115-19 60121-19 60119-11 60118-19 60110-11 60115-11 60118-S 60111-11 60120-S 60116-25 60119-S 60111-S 60115-S 60121-25 60120-25 60120-11 60120-19 60122-19 60118-25 60122-25 60121-S 60116-19 60110-19 60122-11 60117-S 60110-S 60110-25 60111-19 60119-19 60118-11 60117-19 60117-11 60111-25 60116-S 60119-25 60121-11 60115-25 60116-11
102092-944PK 201.43 USD InStock
102092-944 102092-900 102092-922 102092-920 102092-942 102092-926 102092-948 102092-904 102092-924 102092-946 102092-902 102092-908 102092-906 102092-928 102092-870 102092-952 102092-930 102092-896 102092-874 102092-950 102092-894 102092-872 102092-956 102092-978 102092-934 102092-912 102092-878 102092-932 102092-954 102092-910 102092-876 102092-898 102092-938 102092-916 102092-914 102092-936 102092-918 102092-882 102092-880 102092-940 102092-884
MicroMounts™ Crystal Manipulation Tool, Electron Microscopy Sciences
Specimen Mounting Supplies
MicroMounts™ are the highest performance tools for manipulating and mounting protein crystals, virus crystals, and small molecule/inorganic crystals, as well as all other small, fragile samples. They are ideal for both conventional and high-throughput X-ray crystallography applications.
  • MicroMounts™ of a thin (7.5 µm thick) micro-fabricated polyimide film, attached to a solid non-magnetic stainless steel pin
  • Film is wrapped around the steel pin; provides excellent stiffness and convenient scoop-like action in retrieving and handling crystals
Film pattern of Crystal Aperture (A) sizes from 10 to 600 µm, with minimal (B) width around the aperture to minimize background scatter in all orientations. Wicking Hole (C) connected via drainage channel (D) to the crystal aperture. Hole size is compatible with size 15 or XF paper wicks. This 'fountain-pen'-like design, allows easier removal of excess liquid, without touching the crystals with the wick. Alignment cross (E) located a fixed distance from the center of the crystal aperture. Allow easy automated alignment of the crystal aperture (A). Crystal aperture size code (F) around the alignment cross, allows automated recognition of Mount design, aperture, and crystal size, which determines boundaries of aperture (to be avoided), required alignment tolerance, ad area of crystal available to be scanned for optimum diffraction. Initial beam size can then be automatically selected. Aperture size (G) in µm can be read directly off the mount. Orientation mark (H) allows the front and back of the mount to be automatically distinguished.
Solid 316 non-magnetic stainless steel pins. The 11, 19, and 25 mm pin lengths, give pin base-to-crystal center distance compatible to those of nylon loops mounted on 10, 18 and 24 mm pins, respectively. This solid pin will not suck up crystal and liquid. Pin is beveled at film end, allows good visibility of the crystals for wide range of pin orientation during crystal retrieval. Pin can be cut (with cutter pliers) to any desired   length, or can be bent to place crystal in desired orientation. Pin is fitted to all standard bases.
In general, the 19 mm or SPINE length pins is recommended. Copper post style goniometer bases and have been using 10 or 11 m pins with them, consider the B3 copper post base which accepts full 19 mm length pins.
Delivery information: Assortment boxes of apertures recommended, amount aperture can be matched to the size of the crystal. Choose pin lengths from 11, 19, 25, or SPIN. Each box contains 20 MicroMount™ with your choice of size and pin length. Small (A1), includes: 10, 20, 30 and 40 µm apertures. Medium (A2), includes: 75, 150, and 200 µm apertures. Large (A3), includes: 300, 400, 500, and 600 µm apertures Individual MicroMount™.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty