Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Goniometer Bases, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60142-06-B3 60140-01-B3A 60142-06-B1 60145-100 60145-20 60142-06-B3A 60140-01-B3 60140-01-B2 60142-06-B3S 60141-03-B3 60143-20-B1 60142-06-B2 60140-01-B1 60140-01-B3S 60140-01-B1A 60141-03-B3A 60141-03-B2 60141-03-B3S 60141-03-B1A 60142-06-B1A
102092-998EA 28.78 USD InStock
102092-998 102093-040 102093-010 102093-042 102093-020 102093-012 102093-022 102093-000 102093-014 102093-024 102093-002 102093-016 102093-026 102093-018 102093-028 102093-006 102093-008 102092-992 102092-996 102092-994
Goniometer Bases, Electron Microscopy Sciences
Specimen Mounting Supplies
Goniometer Bases tightly capture the MicroRT™ tubing to allow seamless room-temperature and low-temperature data collection. Available in several different base styles that are compatible with MicroRT™ tubing and with standard cryo-tools, cryo-vials, magnetic goniometer head mounts, and most automating hardware.
  • Base B1 is available in two versions: B1 (with flat ledge) and B1A (without the ledge, for ALS-style grippers used at ALS, NSLS and CHESS)
  • Base B2 is a copper post-style base that accepts 11/12 mm MicroMounts™. Available only with the flat edge
  • Base B3 is available in three versions: B3 for 19 mm MicroMounts™, with the flat ledge; B3A for 19 mm MicroMount™, without the ledge; and B3S for SPINE length MicroMount™, without ledge
  • B4 is a 3 mm diameter “brass pin” style base, with the smaller central hole to tightly hold MicroMount™ and an extension to capture MicroRT™ tubing
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty