Would you like to log in again?
Stage Micrometer Model SM-12, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60210-12PG 60210-12PGC
102680-922EA 406.94 USD InStock
102680-922 102093-102
Stage Micrometer Model SM-12, Electron Microscopy Sciences
Micrometers
Linear Reference Scales are available in Chrome-on-Glass (G) and Chrome-on-Opal glass (O) or Chrome-on Quartz (Q)
Positive image are mostly clear or white.
Ordering information: The Opal and Quartz are offered as a special only and must be requested.
Packaging: All EMS micrometers come in a sturdy wooden box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty