Would you like to log in again?
IAM-5 Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60216-5MNGC 60216-5SNGC 60216-5LNG 60216-5SNG 60216-5MNG 60216-5LNGC
102093-138EA 1582.72 USD InStock
102093-138 102093-140 102093-142 102093-144 102093-146 102093-136
IAM-5 Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
This ideal standard for verifying or qualifying multiple image analysis parameters such as optical distortion, alignment between systems or optical paths, and cell areas for particle counting.
The image area is 20×50 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty