Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Model IAM-MET, Grain Size/Nodularity Analysis Standard, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

60216-METC
102093-152EA 1240.77 USD InStock
102093-152
Model IAM-MET, Grain Size/Nodularity Analysis Standard, Electron Microscopy Sciences
Standards Color Reference and Spectroscopy Standards
The IAM-MET is a multi-morphology calibration standard designed specifically for ASTM.

  • Image: Chrome on glass/positive. Calibrated 32 points.
  • Overall Size: 76.2 mm x 25.4 mm (3” x 1”)

The IAM-MET has all plates as found in IAM-1 except for plate 4. This plate contains 9 images of irregular shapes, simulating features commonly found in metallurgical applications.

Certifications: As required by the ASTM standard, the IAM-MET comes with secondary certification of plates 1, 2 and 8.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty