Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, May 25. 
Standard Reticles with Single Cross Line, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60252-21 60251-19 60251-21 60250-21 60252-19 60250-19
102093-160EA 268.34 USD InStock
102093-160 102093-162 102093-164 102093-166 102093-168 102093-158
Standard Reticles with Single Cross Line, Electron Microscopy Sciences
Graticules Reticles and Grids
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty