Would you like to log in again?
Standard Reticles with Scales, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60272-19 60271-19 60273-19 60273-21 60270-21 60274-21 60272-21 60276-19 60271-21 60276-21 60274-19 60270-19
102093-216EA 268.34 USD InStock
102093-216 102093-226 102093-218 102093-228 102093-206 102093-208 102093-210 102093-220 102093-212 102093-222 102093-214 102093-224
Standard Reticles with Scales, Electron Microscopy Sciences
Graticules, Reticles and Grids
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty