Would you like to log in again?
Citrajet® Low Foaming Liquid Acid Cleaners, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Citrajet®
60532-04 60532-01
102093-290EA 176.37 USD InStock
102093-290 102093-292
Citrajet® Low Foaming Liquid Acid Cleaners, Electron Microscopy Sciences
Cleaning Agents Detergents and Surface Cleansers
Low foaming, phosphate-free, concentrated, liquid cleaner and metal brightener for high performance removal of metal oxides, scale, salts and inorganic residues in lab-ware washers, parts washers, power-spray systems and manual and ultrasonic cleaning.
  • Ideal for acid washing and rinsing
  • Dilute 2:100 and it has a pH 2.5
Corrosion inhibited and free rinsing leaving no interfering residues.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty