Would you like to log in again?
Detergent 8® Low Foaming Phosphate Free Detergents , Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Detergent 8®
60538-04 60538-01
102093-304CS 928.38 USD InStock
102093-304 102093-302
Detergent 8® Low Foaming Phosphate Free Detergents , Electron Microscopy Sciences
Cleaning Agents Detergents and Surface Cleansers
Non-ionic, concentrated detergents for labware washers, PC board washers, power-spray systems, ultrasonic cleaning systems and manual cleaning.
  • Dilute 3:100 and it has a pH 11
Specially formulated to remove oils, resin rosins and fluxes from printed circuit board, electronic part, phosphate sensitive labware and nuclear contaminated equipment with no interfering conductive residues. Excellent replacement for solvent cleaning.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty