Would you like to log in again?
EMS Bell Jar Shine, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60540-01
102093-310EA 31.66 USD InStock
102093-310
EMS Bell Jar Shine, Electron Microscopy Sciences
Cleaning Agents Detergents and Surface Cleansers
This unique cleaning compound will not only clean but it polishes as well. Meant to be used on any surface which is either glass, metal, porcelain, ceramic and even chrome surfaces. Perfect for the cleaning of your bell jars from carbon and sputter coaters, as well as any surfaces in the lab that are metal, ceramic, steel etc.
Delivery information: Supplied in a 1 lb container.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty