Would you like to log in again?
Invisible Glass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60811-01
102093-318EA 14.13 USD InStock
102093-318
Invisible Glass, Electron Microscopy Sciences
Cleaning Agents Detergents and Surface Cleansers
Specially formulated, Invisible Glass quickly and easily removes dust, dirt, oil, grease, fingerprints, smoke film, haze and more from glass surfaces, such as windows, mirrors, computer screens, display cabinets, and photocopiers.
Invisible Glass contains no water and soaps, surfactants or foams so it will leave no residue that will either streak or haze.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty