Would you like to log in again?
Multi-Microplate Genie®, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61042-10 61042-12
102093-398EA 1616.56 USD InStock
102093-398 102093-400
Multi-Microplate Genie®, Electron Microscopy Sciences
Shakers and Mixers Microplate Shakers
Microplate Mixer
  • Aggressive mixing speed from 750-3200 RPM for complete and uniform mixing in any microplate format
  • May be used to shake up to 4 microplates, an optional accessory tray is available to double capacity and shake up to 8 microplates at one time
  • Single capacity microplate mixer is also available
  • Small vortexing orbit of 1.0mm for thorough mixing regardless of sample viscosity
  • Speed and orbit combine to offer true vortexing action in each well of a microplate
  • Accepts most microplate types within the recommendations of the Society for Biomolecular Screening (SBS), even in 384-well formats
  • Built-in timer operates from 1 - 60 minutes or continuous for hands-free use
  • May be used in cold rooms and incubators for temperature sensitive applications
Additional accessories for the Multi-Microplate Genie® available are listed with Microplate Genie®.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty