Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Diagnostic Specimen Mailing Systems, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61510-70 61510-72 61510-71T 61510-74T 61510-73 61510-76
102093-414KT 66.17 USD InStock
102093-414 102093-416 102093-418 102093-420 102093-412 102093-422
Diagnostic Specimen Mailing Systems, Electron Microscopy Sciences
Mailers and Shippers
These ready-to-use combination/triple packaging kit, have been tested and certified for transport of diagnostic shipments as defined by the applicable regulations, in compliance with IATA Packaging Instruction 650, US, Postal Service and DOT regulations.
Each kit includes approved 10 mL specimen tubes, leak-resistant foam shippers that secure and protect 95 kPa-certified specimen tubes, absorbent material - sufficient to absorb complete contents of approved primary receptacles filed with blood, waterproof tape, leak-proof pressure-lock poly bag, corrugated shipping carton, UN3373 Biological Substance label, and step-by-step instruction sheet.
Model 472 outer dimensions: 10 1/2 x 5 5/8 x 3”.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty