Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Mini Mailers and Mailing Sleeves, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61522-34 61523-72 61520-12 61522-95 61523-96 61523-03 61522-04 61523-92 61522-35 61523-02 61522-02 61523-42 61520-40 61521-44 61523-36 61523-41 61521-43 61522-01
102093-492PK 28.06 USD InStock
102093-492 102093-494 102093-482 102093-496 102093-484 102093-520 102093-486 102093-500 102093-522 102093-488 102093-502 102093-512 102093-504 102093-514 102093-506 102093-516 102093-508 102093-518
Mini Mailers and Mailing Sleeves, Electron Microscopy Sciences
Mailers and Shippers
Mini mailers either for temperature sensitive specimens or for non-hazardous laboratory specimens.
Mini Mailers for Temperature Sensitive Shipment:
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents
Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants
Insulating, thick wall polystyrene foam construction
Walls on model 440 are 1” thick; walls on model 312 are 1 ½” thick
Model 312 is in corrugated carton
Standard Mini Mailers:
Maximum breakage protection for non-hazardous laboratory specimens. Ideal for biological, environmental, research and development, public health, industries hygiene, and veterinary use
Accommodate tubes, vials, bottles, petri dishes and cans
Lightweight polystyrene foam construction saves postage
Reusable and recyclable
Standard mailing sleeves available
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty