Would you like to log in again?
U-tek® Ice Packets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61533-06 61533-05
102680-986EA 25.87 USD InStock
102680-986 102093-562
U-tek® Ice Packets, Electron Microscopy Sciences
Cooling Packs
U-tek® Ice Packets are cushion fragile shipments which won’t dilute ice baths or damage samples
The packets may be refrozen and reuse, withstands pressure of 60 lb per square inch for 5 minutes or longer. The refrigerant contains neutral pH distilled deionized water and proprietary materials certified by the US Food and Drug Administration as food grade chemicals.
-1°C / +30°F
They remain frozen longer than regular ice without the melting mess.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty