Would you like to log in again?
96-Well Reversible Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61602-G 61602-Y 61602-N 61602-P 61602-O 61602-A 61602-B
102093-656PK 98.26 USD InStock
102093-656 102093-658 102093-646 102093-648 102093-650 102093-652 102093-654
96-Well Reversible Rack, Electron Microscopy Sciences
Racks Tube Racks
Polypropylene rack has 96 wells to accommodate 0.5 mL micro centrifuge tubes on one side, or 96 wells for 1.5 to 2.0 mL micro centrifuge tubes on the other side. Arranged in standard 8 x 12 array. Handy lid snaps into place in the tabs on each end of rack. Grid lines are molded on the side of the rack that accommodates the smaller tubes making it easier to see the wells. Raised rings are molded on the opposite side of the rack making it easier to replace the 1.5 to 2.0 mL tubes from those wells. Rack measures 9.75 x 4.75 x 1.875".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty