Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
NovaRack and NovaChamber, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61611-4 61610-50 61610-5 61611-1 61611-3 61610-4 61610-3
102093-688EA 20.46 USD InStock
102093-688 102093-690 102093-694 102093-682 102093-696 102093-684 102093-686
NovaRack and NovaChamber, Electron Microscopy Sciences
Racks Cryogenic Vial Racks
Unique Cone-Shaped Storage Racks For Easy Sample Viewing. Easily view, organize and track up to 46 tubes including 24 - 0.5 to 2.0 ml tubes, 14 tubes up to 8ml in size including cryovials and 12x75mm culture/blood collection tubes, 7 - 15 ml tubes and 1 - 50 ml tube. Constructed of durable polypropylene and autoclavable. Use the Stacking Adapter to double available bench space. Turn the rack over and fill with water for freezing. A non-slip rubber bottom (included with each NovaChamber) keeps the water/ice contained for a portable ice chamber rack. For easy sample tracking each rack rack has molded numbers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty