Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Cardboard Freezer Boxes and Dividers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61615-90 61615-75 61615-70 61615-85 61615-65 61615-80
102093-722PK 85.35 USD InStock
102093-722 102093-724 102093-726 102093-728 102093-720 102093-730
Cardboard Freezer Boxes and Dividers, Electron Microscopy Sciences
Boxes Cooling/Freezing Boxes and Bags
For use with stainless steel freezer racks. Store cryogenic vials in liquid nitrogen or in mechanical freezers in our freezer storage boxes.
  • Box footprint is 133×133 mm
Select a box and a partition that ft the size and number of vials for your needs. Cardboard boxes can be labelled with Cryo-Tags for easy identification.
Packaging: Both the boxes and dividers are available in packs of 12.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty