Would you like to log in again?
CoolSafe Box and CoolBag, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CoolSafe™
61953-01
102093-848EA 55.08 USD InStock
102093-848
CoolSafe Box and CoolBag, Electron Microscopy Sciences
Boxes Cooling/Freezing Boxes and Bags
The CoolSafe box is made of triple density expanded polystyrene and is molded under extreme pressure to create a durable container that guarantees temperature stability.
  • The CoolBag is an insulated cooler for added coolness of critical samples
Use the shoulder strap for easy transporting from lab to lab.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty