Would you like to log in again?
Frigimat Dry Ice Maker, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Frigimat®
62056-10 62056-22 62056-20
102093-912EA 2623.56 USD InStock
102093-912 102093-914 102093-916
Frigimat Dry Ice Maker, Electron Microscopy Sciences
Ice Makers
The Frigimat® Dry Ice Maker eliminates the need to buy large quantities of dry ice when only a few blocks are needed. It employs the Joule-Thomson effect to produce a solid 454 g (16 oz.) block of dry ice in one to two minutes.
  • A brass relief valve safely vents excess gas. Removing the dry ice block requires just 30 seconds
  • Each 50 lbs. cylinder of liquid CO2 will make 8 to 9 blocks of dry ice, each measuring 8.5cm (3 1/8") x 11cm (4 3/8")
  • Unit is made from epoxy-coated steel with an aluminum ice chamber and brass valves with four non-skid rubber feet
Size: 300x170x130 mm (12x6.5x5")
Shipping weight: 5.5 kg (12 lbs.)
Ordering information: Includes a 1.8m (6 ft) nickel-plated copper connecting tube with standard GCA 320 (0.830" – 14) thread couplings for easy attachment to an LCO2 cylinder.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty