Would you like to log in again?
Frigimat Cub Dry Ice Maker, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62057-10
102093-918EA 1049.43 USD InStock
102093-918
Frigimat Cub Dry Ice Maker, Electron Microscopy Sciences
Ice Makers
Economical block dry ice maker for on-demand use. In only a few minutes, this compact device can be attached to a liquid CO2 cylinder and will produce a ready-to-use 250 - 300 g block of dry ice.
  • Each 50 lbs. cylinder makes from 10 to 16 blocks of dy ice
  • Set-up and use is safe and simple
  • Dry ice production can be viewed through the transparent polycarbonate body that secures without clamps or screws or tools ensuring easy removal of the solid block of dry ice and connection from the CO2 cylinder
  • A rugged nickel plated “pigtail” connects the Frigimat® Cub – Dry Ice Maker to the CO2 cylinder valve
  • The system operates at very low pressure (displayed on the built-in dial gauge) and incorporates multiple pressure relief safeguards
Cylinder Connection: 1.8 m (6 ft.) nickel-plated copper connecting tube with standard CGA 320 thread coupling for easy attachment to a CO2 cylinder.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty