Would you like to log in again?
Nylon Biopsy Bag, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62324-75 62324-98 62324-35
102094-014PK 127.22 USD InStock
102094-014 100490-158 102094-016
Nylon Biopsy Bag, Electron Microscopy Sciences
Bags Sample Bags
Biopsy bags are made from nylon cloth with 0.2 mm mesh diameter.
  • All seams are heat-sealed
  • Reduces the loss of small biopsy specimens
  • Reduces cross contamination and pressure artifacts by using sponges
This allows for unrestricted fluid movement around the tissue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty