Would you like to log in again?
Strip Disposable Mold, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62351-37 62351-30
102094-024PK 99.54 USD InStock
102094-024 102094-028
Strip Disposable Mold, Electron Microscopy Sciences
Moulds Moulds for Histology
Strip disposable molds are manufactured from high impact polystyrene, offering a low cost alternative to stainless steel
These molds are available in a choice of sizes and depths. The molds have a smooth interior finish and rounded corners for ease of removal. There are five molds per strip and they are packed 100 strips to a case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty