Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Simport CytoSep™ Single Cytology Funnels, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62750-45 62750-30 62750-35 62750-20 62750-40 62750-10
102094-422PK 116.47 USD InStock
102094-422 102094-420 102094-430 102094-428 102094-426 102094-424
Simport CytoSep™ Single Cytology Funnels, Electron Microscopy Sciences
Funnels Cytology Funnels
Fully compatible with the Shandon CytoSpin® Centrifuge, the Simport CytoSep™ Cytology Funnels can be used to deposit a thin layer of cells in a clearly defined area of a microscope slide. The filter card absorbs any access fluid. These Cytology Funnels have the filter cards pre-attached for consistent, reliable results. No alignment necessary. All disposable funnels are packaged with closure caps to seal in specimen for added protection. All components are also available separately.
For sample volumes of up to 0.5 mL, use the Single CytoSep™ Cytology Funnel with a White Filter Card & Cap. It provides a cell deposition area of 6 mm (28 mm squares) and allows one sample to be run simultaneously on a single slide. It is not necessary to use a different stainless steel slide clip.
For sample volumes of up to 0.4 mL, such as spinal Fluids, use the Single CytoSep™ Cytology Funnel with Brown Filter Card & Cap. It allows for a slower absorption of fluids. All the individual components are also available separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty