Would you like to log in again?
Oven Incubator, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
63230-20 63230-10
102094-554EA 2516.44 USD InStock
102094-554 100498-358
Oven Incubator, Electron Microscopy Sciences
Incubators Standard Incubators
This Oven/Incubator with aluminum inner chamber and venting is up to the task of providing many years of long service in busy labs.
  • Oven works on the gravity convection heating method
  • An aluminum chamber provides excellent temperature uniformity
  • A thermometer port at the top of the oven accommodates any partial immersion thermometer
  • Heats up rapidly (56°C in 9 minutes)
  • The chamber is vented through the top to allow for the removal of moisture and other vapors
  • The unit comes complete with one removable metal shelf, three-wire cord, and plug
  • Use for general drying and incubation applications and polymerization of resins
The cast aluminum chamber provides for dependable temperature uniformity and the vent efficiently expels moisture and vapors. The unit comes complete with a removable shelf.
Overall Dimensions: HxWxD: 225 x185 x135 mm
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty